Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CETN2
Entrez Gene ID
102172877
Gene Name
centrin 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_017189740: 167251 - 171587 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005700491.3 XP_005700548.2 3 - 4 413 109
XM_013976312.2 XP_013831766.1 3 - 4 411 109
GenBank mRNA JO397552.2 - 2 - 3 342 109
JO595664.2 - 3 - 4 477 109

View Full Product Details