Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BTLA
Entrez Gene ID
102181629
Gene Name
B and T lymphocyte associated
Gene Aliases
-
Chromosome Location
Chr.1: 56978310 - 57012771 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005674919.3 XP_005674976.1 2 - 3 477 69

View Full Product Details