Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DYTN
Entrez Gene ID
102174885
Gene Name
dystrotelin
Gene Aliases
-
Chromosome Location
Chr.2: 40922894 - 40981625 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018066539.1 XP_017922028.1 10 - 11 1181 94

View Full Product Details