Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRPL33
Entrez Gene ID
102173666
Gene Name
mitochondrial ribosomal protein L33
Gene Aliases
-
Chromosome Location
Chr.11: 71652998 - 71662441 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005686871.3 XP_005686928.1 3 - 4 237 103
GenBank mRNA KA338500.1 - 1 - 2 289 103

View Full Product Details