Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC102171800
Entrez Gene ID
102171800
Gene Name
signal-regulatory protein beta-1
Gene Aliases
SIRPB1, SIRPD
Chromosome Location
Chr.13: 58963405 - 58973584 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005688326.3 XP_005688383.2 4 - 5 1176 91

View Full Product Details