Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LY6G6F
Entrez Gene ID
102179719
Gene Name
lymphocyte antigen 6 family member G6F
Gene Aliases
-
Chromosome Location
Chr.23: 22358563 - 22364611 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005696664.2 XP_005696721.2 1 - 2 97 78

View Full Product Details