Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- KCNMB3
- Entrez Gene ID
-
102168667
- Gene Name
- potassium calcium-activated channel subfamily M regulatory beta subunit 3
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 87446105 - 87551930 on Build ARS1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JO596546.2 |
- |
1 - 2 |
96 |
102 |
View Full Product Details