Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSC22D3
Entrez Gene ID
102174369
Gene Name
TSC22 domain family member 3
Gene Aliases
-
Chromosome Location
Chr.Un|NW_017190149: 12795 - 80640 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018045120.1 XP_017900609.1 2 - 3 608 75
XM_018045121.1 XP_017900610.1 2 - 3 618 75
XM_018045125.1 XP_017900614.1 2 - 3 491 75

View Full Product Details