Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AQP6
Entrez Gene ID
102172329
Gene Name
aquaporin 6
Gene Aliases
-
Chromosome Location
Chr.5: 29552477 - 29556318 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005679981.3 XP_005680038.1 2 - 3 671 80

View Full Product Details