Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TCF21
Entrez Gene ID
102175768
Gene Name
transcription factor 21
Gene Aliases
-
Chromosome Location
Chr.9: 59157638 - 59160659 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005684779.3 XP_005684836.1 1 - 2 731 87

View Full Product Details