Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TOR1B
Entrez Gene ID
102173310
Gene Name
torsin family 1 member B
Gene Aliases
-
Chromosome Location
Chr.11: 99667062 - 99676517 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005687268.3 XP_005687325.2 2 - 3 552 64
XM_018055958.1 XP_017911447.1 2 - 3 534 64
XM_018055959.1 XP_017911448.1 2 - 3 534 64

View Full Product Details