Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CENPBD1
Entrez Gene ID
102171182
Gene Name
CENPB DNA-binding domain containing 1
Gene Aliases
-
Chromosome Location
Chr.18: 16137776 - 16141182 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005691940.3 XP_005691997.1 1 - 1 1856 70

View Full Product Details