Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KRT38
Entrez Gene ID
102175892
Gene Name
keratin 38
Gene Aliases
-
Chromosome Location
Chr.19: 41293031 - 41299795 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005693812.3 XP_005693869.2 2 - 3 1828 105

View Full Product Details