Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TCHH
Entrez Gene ID
102188454
Gene Name
trichohyalin
Gene Aliases
THH, THL, TRHY
Chromosome Location
Chr.3: 101934398 - 101942370 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018046108.1 XP_017901597.1 1 - 2 166 104
XM_018046109.1 XP_017901598.1 2 - 3 182 104
XM_018046110.1 XP_017901599.1 2 - 3 182 104
XM_018046111.1 XP_017901600.1 2 - 3 182 104
XM_018046112.1 XP_017901601.1 2 - 3 183 104
XM_018046113.1 XP_017901602.1 2 - 3 183 104
XM_018046114.1 XP_017901603.1 1 - 2 181 104
XM_018046115.1 XP_017901604.1 2 - 3 183 104
XM_018046116.1 XP_017901605.1 2 - 3 184 104
XM_018046117.1 XP_017901606.1 2 - 3 182 104
GenBank mRNA KA336006.1 - 1 - 2 1197 104

View Full Product Details