Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HEATR4
Entrez Gene ID
102184157
Gene Name
HEAT repeat containing 4
Gene Aliases
-
Chromosome Location
Chr.10: 18150711 - 18188440 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013967282.2 XP_013822736.2 3 - 4 2086 58
XM_018053995.1 XP_017909484.1 3 - 4 2086 58

View Full Product Details