Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KAT14
Entrez Gene ID
102189321
Gene Name
lysine acetyltransferase 14
Gene Aliases
-
Chromosome Location
Chr.13: 37781039 - 37807839 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013968598.2 XP_013824052.2 4 - 5 744 85

View Full Product Details