Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ANKFN1
Entrez Gene ID
102183360
Gene Name
ankyrin repeat and fibronectin type III domain containing 1
Gene Aliases
-
Chromosome Location
Chr.19: 6727359 - 6887602 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013971777.2 XP_013827231.2 4 - 5 659 86

View Full Product Details