Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNK4
Entrez Gene ID
102190201
Gene Name
potassium two pore domain channel subfamily K member 4
Gene Aliases
-
Chromosome Location
Chr.29: 43016685 - 43022806 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018043594.1 XP_017899083.1 3 - 4 713 77

View Full Product Details