Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAX3
Entrez Gene ID
102183925
Gene Name
paired box 3
Gene Aliases
-
Chromosome Location
Chr.2: 25335604 - 25439167 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005676604.3 XP_005676661.1 4 - 5 973 104

View Full Product Details