Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
REG4
Entrez Gene ID
102173903
Gene Name
regenerating family member 4
Gene Aliases
-
Chromosome Location
Chr.3: 97053865 - 97080491 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005677782.3 XP_005677839.1 4 - 5 447 90

View Full Product Details