Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IL26
Entrez Gene ID
102176200
Gene Name
interleukin 26
Gene Aliases
-
Chromosome Location
Chr.5: 44912744 - 44927565 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005680207.2 XP_005680264.2 4 - 5 536 103

View Full Product Details