Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYTL1
Entrez Gene ID
102190869
Gene Name
cytokine like 1
Gene Aliases
-
Chromosome Location
Chr.6: 104454473 - 104459757 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005681894.3 XP_005681951.1 1 - 2 564 77

View Full Product Details