Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EMSY
Entrez Gene ID
102175878
Gene Name
EMSY transcriptional repressor, BRCA2 interacting
Gene Aliases
-
Chromosome Location
Chr.15: 26990509 - 27072511 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005690007.3 XP_005690064.1 10 - 11 1799 97
XM_005690008.3 XP_005690065.1 10 - 11 1798 97
XM_013969807.2 XP_013825261.1 9 - 10 1757 97
XM_013969808.2 XP_013825262.1 9 - 10 1646 97
XM_013969809.2 XP_013825263.1 10 - 11 1800 97
XM_013969810.2 XP_013825264.1 9 - 10 1645 97
XM_013969811.2 XP_013825265.1 10 - 11 1798 97
XM_013969812.2 XP_013825266.1 10 - 11 1800 97
XM_018059320.1 XP_017914809.1 9 - 10 1699 97
GenBank mRNA JO559705.2 - 9 - 10 1588 97

View Full Product Details