Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EMSY
Entrez Gene ID
102175878
Gene Name
EMSY transcriptional repressor, BRCA2 interacting
Gene Aliases
-
Chromosome Location
Chr.15: 26990509 - 27072511 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005690007.3 XP_005690064.1 17 - 18 2960 96
XM_013969807.2 XP_013825261.1 16 - 17 2918 96
XM_013969808.2 XP_013825262.1 16 - 17 2807 96
XM_013969809.2 XP_013825263.1 16 - 17 2796 96
XM_013969812.2 XP_013825266.1 17 - 18 2961 96
XM_018059320.1 XP_017914809.1 16 - 17 2860 96
GenBank mRNA JT708455.1 - 15 - 16 2549 96

View Full Product Details