Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC102169746
Entrez Gene ID
102169746
Gene Name
zinc finger protein 501
Gene Aliases
ZFP92
Chromosome Location
Chr.Un|NW_017214957: 55353 - 58314 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005700553.2 XP_005700610.2 2 - 3 157 78

View Full Product Details