Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYNC
Entrez Gene ID
102170923
Gene Name
syncoilin, intermediate filament protein
Gene Aliases
-
Chromosome Location
Chr.2: 14738980 - 14759735 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018057857.1 XP_017913346.1 3 - 4 1446 96

View Full Product Details