Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
ASIP
Entrez Gene ID
100860915
Gene Name
agouti signaling protein
Gene Aliases
agouti
Chromosome Location
Chr.13: 63228708 - 63249541 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018057735.1 XP_017913224.1 2 - 3 272 77
XM_018057736.1 XP_017913225.1 2 - 3 221 77
GenBank mRNA GU076166.2 - 1 - 2 31 77
KA320319.1 - - 453 77
KT247425.1 - 1 - 2 166 77

View Full Product Details