Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC102171498
Entrez Gene ID
102171498
Gene Name
spermidine/spermine N(1)-acetyltransferase-like protein 1
Gene Aliases
SATL1
Chromosome Location
Chr.29: 39561965 - 39616008 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005700624.2 XP_005700681.2 3 - 4 307 89

View Full Product Details