Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LYSMD1
Entrez Gene ID
102176928
Gene Name
LysM domain containing 1
Gene Aliases
-
Chromosome Location
Chr.3: 100503931 - 100511947 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005677626.3 XP_005677683.1 2 - 3 1168 67
GenBank mRNA JT686457.1 - 5 - 6 1073 67

View Full Product Details