Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NEFL
Entrez Gene ID
102179945
Gene Name
neurofilament light
Gene Aliases
-
Chromosome Location
Chr.8: 71973603 - 71979306 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005684017.3 XP_005684074.2 2 - 3 1311 96

View Full Product Details