Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PI15
Entrez Gene ID
102174675
Gene Name
peptidase inhibitor 15
Gene Aliases
-
Chromosome Location
Chr.14: 43894711 - 43930815 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005689105.3 XP_005689162.1 5 - 6 850 79
XM_018058392.1 XP_017913881.1 5 - 6 877 79

View Full Product Details