Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VSIR
Entrez Gene ID
102175186
Gene Name
V-set immunoregulatory receptor
Gene Aliases
C28H10orf54
Chromosome Location
Chr.28: 17703220 - 17734425 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005699167.3 XP_005699224.1 1 - 2 245 64

View Full Product Details