Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSC22D1
Entrez Gene ID
102178181
Gene Name
TSC22 domain family member 1
Gene Aliases
-
Chromosome Location
Chr.12: 71744971 - 71867245 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018056767.1 XP_017912256.1 1 - 2 3031 108

View Full Product Details