Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM71
Entrez Gene ID
102171555
Gene Name
tripartite motif containing 71
Gene Aliases
-
Chromosome Location
Chr.22: 7097747 - 7159684 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018066746.1 XP_017922235.1 3 - 4 1230 83

View Full Product Details