Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HEPACAM
Entrez Gene ID
102182282
Gene Name
hepatic and glial cell adhesion molecule
Gene Aliases
-
Chromosome Location
Chr.29: 27705874 - 27724660 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018043578.1 XP_017899067.1 5 - 6 1262 62
GenBank mRNA JO632295.2 - 5 - 6 1294 62

View Full Product Details