Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HENMT1
Entrez Gene ID
102187549
Gene Name
HEN methyltransferase 1
Gene Aliases
-
Chromosome Location
Chr.3: 85844768 - 85855688 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005678010.3 XP_005678067.2 1 - 2 105 81

View Full Product Details