Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD70
Entrez Gene ID
102188305
Gene Name
CD70 molecule
Gene Aliases
-
Chromosome Location
Chr.7: 92585991 - 92590909 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005682534.3 XP_005682591.2 2 - 3 388 95

View Full Product Details