Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
REM2
Entrez Gene ID
102183795
Gene Name
RRAD and GEM like GTPase 2
Gene Aliases
-
Chromosome Location
Chr.10: 79474838 - 79482328 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018054571.1 XP_017910060.1 2 - 3 2162 104
XM_018054572.1 XP_017910061.1 3 - 4 4132 104
XM_018054573.1 XP_017910062.1 3 - 4 1946 104
XM_018054574.1 XP_017910063.1 4 - 5 3916 104

View Full Product Details