Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C11H9orf78
Entrez Gene ID
102182416
Gene Name
chromosome 11 C9orf78 homolog
Gene Aliases
-
Chromosome Location
Chr.11: 99685549 - 99694190 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005687300.3 XP_005687357.2 5 - 6 420 81
XM_018055961.1 XP_017911450.1 4 - 5 349 81

View Full Product Details