Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMCR8
Entrez Gene ID
102191734
Gene Name
SMCR8-C9orf72 complex subunit
Gene Aliases
-
Chromosome Location
Chr.19: 34012104 - 34021029 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005693612.3 XP_005693669.2 1 - 2 2855 90
GenBank mRNA KA348535.1 - 1 - 2 3733 90

View Full Product Details