Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MCCD1
Entrez Gene ID
102187820
Gene Name
mitochondrial coiled-coil domain 1
Gene Aliases
-
Chromosome Location
Chr.23: 22204299 - 22205984 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005696609.3 XP_005696666.1 1 - 2 419 65
GenBank mRNA JT674978.1 - 2 - 3 461 65

View Full Product Details