Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBFA
Entrez Gene ID
102183557
Gene Name
ribosome binding factor A
Gene Aliases
-
Chromosome Location
Chr.24: 365523 - 379989 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005696938.3 XP_005696995.1 6 - 7 742 104
XM_005696939.3 XP_005696996.1 6 - 7 744 104

View Full Product Details