Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACRV1
Entrez Gene ID
102185536
Gene Name
acrosomal vesicle protein 1
Gene Aliases
-
Chromosome Location
Chr.29: 28401291 - 28407744 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005699596.2 XP_005699653.2 1 - 2 139 95
XM_005699597.2 XP_005699654.2 1 - 2 54 95
XM_005699598.2 XP_005699655.2 1 - 2 54 95

View Full Product Details