Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
KLHL30
Entrez Gene ID
106501991
Gene Name
kelch like family member 30
Gene Aliases
-
Chromosome Location
Chr.3: 3246836 - 3258905 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018040298.1 XP_017895787.1 4 - 5 1134 56
GenBank mRNA JO356437.2 - 2 - 3 175 56
JT755988.1 - 2 - 3 174 56

View Full Product Details