Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRC31
Entrez Gene ID
102190076
Gene Name
leucine rich repeat containing 31
Gene Aliases
-
Chromosome Location
Chr.1: 97036909 - 97064067 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013964434.2 XP_013819888.2 2 - 3 255 96

View Full Product Details