Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ENTHD1
Entrez Gene ID
102185991
Gene Name
ENTH domain containing 1
Gene Aliases
-
Chromosome Location
Chr.5: 110089924 - 110171655 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005681180.2 XP_005681237.2 3 - 4 705 75

View Full Product Details