Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRXCR2
Entrez Gene ID
102191327
Gene Name
glutaredoxin and cysteine rich domain containing 2
Gene Aliases
-
Chromosome Location
Chr.7: 53372824 - 53392465 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005683092.3 XP_005683149.1 2 - 3 555 91

View Full Product Details