Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ISCA2
Entrez Gene ID
102190699
Gene Name
iron-sulfur cluster assembly 2
Gene Aliases
-
Chromosome Location
Chr.10: 17335140 - 17337599 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005686079.3 XP_005686136.1 3 - 4 306 106
XM_005686080.3 XP_005686137.1 3 - 4 303 106

View Full Product Details