Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC102189740
Entrez Gene ID
102189740
Gene Name
solute carrier family 22 member 10
Gene Aliases
SLC22A10
Chromosome Location
Chr.29: 41797562 - 41884197 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005699807.3 XP_005699864.2 7 - 8 1146 85

View Full Product Details