Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAM83F
Entrez Gene ID
102186284
Gene Name
family with sequence similarity 83 member F
Gene Aliases
-
Chromosome Location
Chr.5: 110277254 - 110314067 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018048797.1 XP_017904286.1 2 - 3 691 62
GenBank mRNA JT662704.1 - 3 - 4 599 62
KA346728.1 - 2 - 3 693 62

View Full Product Details